Learn More
TA Instruments Stainless Steel, 20 mm, 0.5 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580848 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (20 mm, 0.5 degree) is an upper cone compatible with lower Peltier plates. It uses Smart Swap 2 geometry to provide secure placement and precise rheological measurements on HR X0 instruments.
Catalog No. 50-005-80848 Supplier TA Instruments Supplier No. 511203.945
HR X0 Stainless Steel Cone (20 mm, 0.5 degree) is an upper cone compatible with lower Peltier plates. It uses Smart Swap 2 geometry to provide secure placement and precise rheological measurements on HR X0 instruments.