Learn More
TA Instruments Stainless Steel, 20 mm, 1 deg Cone, NST, HR X0
Catalog No. 5000580800 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0 Stainless Steel Upper Cone (20 mm, 1 deg, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only.
Catalog No. 50-005-80800 Supplier TA Instruments Supplier No. 511204.946
Waters TA Instruments HR X0 Stainless Steel Upper Cone (20 mm, 1 deg, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only.