Learn More
TA Instruments Stainless Steel, 20 mm, 1 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580626 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (20 mm, 1 degree) is designed for lower Peltier plates and uses Smart Swap 2 geometry for accurate placement and reliable sample measurement on HR X0 instruments.
Catalog No. 50-005-80626 Supplier TA Instruments Supplier No. 511204.945
HR X0 Stainless Steel Cone (20 mm, 1 degree) is designed for lower Peltier plates and uses Smart Swap 2 geometry for accurate placement and reliable sample measurement on HR X0 instruments.