Learn More
TA Instruments Stainless Steel, 20 mm, 2 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580864 Shop All Waters TA Instruments Products
Encompass_Preferred
DH X0 Stainless Steel Cone (20 mm, 2 degree) is an upper cone for lower Peltier plates. It features Smart Swap 2 geometry for precise placement and accurate rheological measurements on DH X0 instruments.
Catalog No. 50-005-80864 Supplier TA Instruments Supplier No. 511206.945
DH X0 Stainless Steel Cone (20 mm, 2 degree) is an upper cone for lower Peltier plates. It features Smart Swap 2 geometry for precise placement and accurate rheological measurements on DH X0 instruments.