Learn More
TA Instruments Stainless Steel, 20 mm, 4 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580844 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (20 mm, 4 degree) is compatible with lower Peltier plates. Smart Swap 2 geometry ensures secure installation and reproducible measurement of samples for HR X0 instruments.
Catalog No. 50-005-80844 Supplier TA Instruments Supplier No. 511207.945
HR X0 Stainless Steel Cone (20 mm, 4 degree) is compatible with lower Peltier plates. Smart Swap 2 geometry ensures secure installation and reproducible measurement of samples for HR X0 instruments.