Learn More
TA Instruments Stainless Steel, 20 mm Plate, NST, HR X0
Catalog No. 5000580653 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0 Stainless Steel Upper Plate (20 mm, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only.
Catalog No. 50-005-80653 Supplier TA Instruments Supplier No. 511200.946
Waters TA Instruments HR X0 Stainless Steel Upper Plate (20 mm, NST) is for use with lower Peltier plate. Smart Swap 2 geometry for use with HR X0 instruments only.