Learn More
TA Instruments Stainless Steel, 20 mm Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580870 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Plate (20 mm) is an upper plate compatible with lower Peltier plates. It uses Smart Swap 2 geometry for secure installation and consistent sample contact on HR X0 instruments.
Catalog No. 50-005-80870 Supplier TA Instruments Supplier No. 511200.945
HR X0 Stainless Steel Plate (20 mm) is an upper plate compatible with lower Peltier plates. It uses Smart Swap 2 geometry for secure installation and consistent sample contact on HR X0 instruments.