Learn More
TA Instruments Stainless Steel, 20 mm, Quick Change Plate, HR X0, DHR, ARES-G2
Catalog No. 5000580749 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2 Stainless Steel Quick Change Plate (20 mm) is compatible with Advanced Peltier Plates and the Standard Flat Plate system. Designed for secure placement, easy installation, and precise rheological measurements across HR X0, DHR, and ARES-G2 platforms.
Catalog No. 50-005-80749 Supplier TA Instruments Supplier No. 402491.901
Waters TA Instruments HR X0, DHR, ARES-G2 Stainless Steel Quick Change Plate (20 mm) is compatible with Advanced Peltier Plates and the Standard Flat Plate system. Designed for secure placement, easy installation, and precise rheological measurements across HR X0, DHR, and ARES-G2 platforms.