Learn More
TA Instruments Stainless Steel, 25 mm, Quick Change Plate, HR X0, DHR, ARES-G2, APS
Catalog No. 5000580818 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Quick Change Plate (25 mm) is intended for use with HR X0 or DHR Advanced Peltier Plates and ARES-G2 Advanced Peltier System with the Standard Flat Plate. Features reliable alignment, secure placement, and reproducible measurement results.
Catalog No. 50-005-80818 Supplier TA Instruments Supplier No. 402624.901
Waters TA Instruments HR X0, DHR, ARES-G2, APS Stainless Steel Quick Change Plate (25 mm) is intended for use with HR X0 or DHR Advanced Peltier Plates and ARES-G2 Advanced Peltier System with the Standard Flat Plate. Features reliable alignment, secure placement, and reproducible measurement results.