Learn More
Alkali Scientific Stainless Steel 3 High Frame for C 500 Canisters
SDP
Catalog No. 5000662041
Encompass_Preferred
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 3 High Frame for C 500 Holds 3 C 500 Canisters Blood Bag Number: BAXTER/FENWAL 4R9955,OrigenCS500, CyroMACS 200 074 402, CharterMed CF 500, FP 500mL and FP FLEX500 Dimensions (D x H x L): 8 11/16 x 18 13/16 x 9/16″ 220 x 478 x 15mm Canister: C 500
Catalog No. 50-006-62041 Supplier Alkali Scientific Supplier No. FC5003
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 3 High Frame for C 500 Holds 3 C 500 Canisters Blood Bag Number: BAXTER/FENWAL 4R9955,OrigenCS500, CyroMACS 200 074 402, CharterMed CF 500, FP 500mL and FP FLEX500 Dimensions (D x H x L): 8 11/16 x 18 13/16 x 9/16″ 220 x 478 x 15mm Canister: C 500