Learn More
Alkali Scientific Stainless Steel 3 High Frame for C 750 Canisters
SDP
Catalog No. 5000662061
Encompass_Preferred
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 3 High Frame for C 750 Holds 3 C 750 Canisters Blood Bag Number: BAXTER/FENWAL 4R9957,OrigenCS750, CyroMACS 200 074 403, CharterMed CF 750, FP 750mL and FP FLEX750 Dimensions (D x H x L): 10 5/16 x 18 13/16 x 9/16″ 262 x 478 x 15mm Canister: C 750
Catalog No. 50-006-62061 Supplier Alkali Scientific Supplier No. FC7503
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 3 High Frame for C 750 Holds 3 C 750 Canisters Blood Bag Number: BAXTER/FENWAL 4R9957,OrigenCS750, CyroMACS 200 074 403, CharterMed CF 750, FP 750mL and FP FLEX750 Dimensions (D x H x L): 10 5/16 x 18 13/16 x 9/16″ 262 x 478 x 15mm Canister: C 750