Learn More
Sigma Aldrich Fine Chemicals Biosciences Stainless steel 304 syring
Catalog No. 502905230
Encompass
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved deflecting tip to prevent coring during needle insertion through the septum.Standard Hub with 304 Stainless Steel Tubing
Catalog No. 50-290-5230 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAD485312EA
304 SS needle with chrome-plated brass Luer hub. Needle is designed with a slightly curved deflecting tip to prevent coring during needle insertion through the septum.Standard Hub with 304 Stainless Steel Tubing