Learn More
Sigma Aldrich Fine Chemicals Biosciences Stainless steel 316 syring1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2612971EA
Encompass
Specifically designed for secure leakproof mating with plastic Luer tip disposable syringes and metal Luer tip glass syringes. female Luer hubs and needles are made of 316 SS. Noncontaminating easy to clean and resistant to corrosion.Micro-Mate Hub with 316 Stainless Steel Tubing
Catalog No. 50-290-0859