Learn More
Alkali Scientific Stainless Steel 4 High Frame for C 1000 Canisters
SDP
Catalog No. 5000662865
Encompass_Preferred
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 4 High Frame for C 1000 Holds 4 C 1000 Canisters Blood Bag Number: BAXTER/FENWAL 4R9959,OrigenCS1000 CyroMACS 200 074 404, CharterMed CF 1000, FP 1000mL and FP FLEX1000 Dimensions (D x H x L): 13 5/16 x 24 15/16 x 9/16″ 338 x 633 x 15mm Canister: C 1000
Catalog No. 50-006-62865 Supplier Alkali Scientific Supplier No. FC10004
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 4 High Frame for C 1000 Holds 4 C 1000 Canisters Blood Bag Number: BAXTER/FENWAL 4R9959,OrigenCS1000 CyroMACS 200 074 404, CharterMed CF 1000, FP 1000mL and FP FLEX1000 Dimensions (D x H x L): 13 5/16 x 24 15/16 x 9/16″ 338 x 633 x 15mm Canister: C 1000