Learn More
Alkali Scientific Stainless Steel 4 High Frame for C 250 Canisters
SDP
Catalog No. 5000663298
Encompass_Preferred
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 4 High Frame for C 250 Holds 4 C 250 Canisters Blood Bag Number: BAXTER/FENWAL 4R9953, OrigenCS250, CyroMACS 200 074 401, CharterMed CF 250, FP 250mL and FP FLEX250 Dimensions (D x H x L): 6 15/16 x 24 15/16 x 9/16″ 176 x 633 x 15mm Canister: C 250
Catalog No. 50-006-63298 Supplier Alkali Scientific Supplier No. FC2504
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 4 High Frame for C 250 Holds 4 C 250 Canisters Blood Bag Number: BAXTER/FENWAL 4R9953, OrigenCS250, CyroMACS 200 074 401, CharterMed CF 250, FP 250mL and FP FLEX250 Dimensions (D x H x L): 6 15/16 x 24 15/16 x 9/16″ 176 x 633 x 15mm Canister: C 250