Learn More
TA Instruments Stainless Steel, 40 mm, 0. 5 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580833 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (40 mm, 0.5 degree) is an upper cone for use with lower Peltier plates. It features Smart Swap 2 geometry for secure and accurate placement, designed exclusively for HR X0 instruments.
Catalog No. 50-005-80833 Supplier TA Instruments Supplier No. 511403.945
HR X0 Stainless Steel Cone (40 mm, 0.5 degree) is an upper cone for use with lower Peltier plates. It features Smart Swap 2 geometry for secure and accurate placement, designed exclusively for HR X0 instruments.