Learn More
TA Instruments Stainless Steel, 40 mm, 1 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580855 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (40 mm, 1 degree) is an upper cone compatible with lower Peltier plates. It uses Smart Swap 2 geometry for precise alignment and is designed for HR X0 instruments only.
Catalog No. 50-005-80855 Supplier TA Instruments Supplier No. 511404.945
HR X0 Stainless Steel Cone (40 mm, 1 degree) is an upper cone compatible with lower Peltier plates. It uses Smart Swap 2 geometry for precise alignment and is designed for HR X0 instruments only.