Learn More
TA Instruments Stainless Steel, 40 mm, 2 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580729 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (40 mm, 2 degree) is an upper cone for use with lower Peltier plates. It features Smart Swap 2 geometry, providing accurate placement and reliable rheological measurements for HR X0 instruments.
Catalog No. 50-005-80729 Supplier TA Instruments Supplier No. 511406.945
HR X0 Stainless Steel Cone (40 mm, 2 degree) is an upper cone for use with lower Peltier plates. It features Smart Swap 2 geometry, providing accurate placement and reliable rheological measurements for HR X0 instruments.