Learn More
TA Instruments Stainless Steel, 40 mm, 4 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580730 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (40 mm, 4 degree) is designed for HR X0 instruments and lower Peltier plates. The Smart Swap 2 geometry ensures secure installation and precise measurement of samples.
Catalog No. 50-005-80730 Supplier TA Instruments Supplier No. 511407.945
HR X0 Stainless Steel Cone (40 mm, 4 degree) is designed for HR X0 instruments and lower Peltier plates. The Smart Swap 2 geometry ensures secure installation and precise measurement of samples.