Learn More
TA Instruments Stainless Steel, 40 mm Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580796 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Plate (40 mm) is an upper plate designed for use with lower Peltier plates. The Smart Swap 2 geometry ensures precise positioning and consistent sample interaction for HR X0 instruments.
Catalog No. 50-005-80796 Supplier TA Instruments Supplier No. 511400.945
HR X0 Stainless Steel Plate (40 mm) is an upper plate designed for use with lower Peltier plates. The Smart Swap 2 geometry ensures precise positioning and consistent sample interaction for HR X0 instruments.