Learn More
TA Instruments Stainless Steel, 50 mm, 1 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580874 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (50 mm, 1 degree) is an upper cone compatible with lower Standard and Advanced Peltier plates. It uses Smart Swap 2 geometry for precise and secure placement on HR X0 instruments.
Catalog No. 50-005-80874 Supplier TA Instruments Supplier No. 511504.945
HR X0 Stainless Steel Cone (50 mm, 1 degree) is an upper cone compatible with lower Standard and Advanced Peltier plates. It uses Smart Swap 2 geometry for precise and secure placement on HR X0 instruments.