Learn More
TA Instruments Stainless Steel, 50 mm, 2 Degree Cone, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580832 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Cone (50 mm, 2 degree) is an upper cone for use with lower Standard and Advanced Peltier plates. It features Smart Swap 2 geometry for accurate alignment and reliable rheological measurements on HR X0 instruments.
Catalog No. 50-005-80832 Supplier TA Instruments Supplier No. 511506.945
HR X0 Stainless Steel Cone (50 mm, 2 degree) is an upper cone for use with lower Standard and Advanced Peltier plates. It features Smart Swap 2 geometry for accurate alignment and reliable rheological measurements on HR X0 instruments.