Learn More
TA Instruments Stainless Steel, 50 mm Plate, Smart Swap 2 Geometry, HR x0 Instruments Only
Catalog No. 5000580782 Shop All Waters TA Instruments Products
Encompass_Preferred
HR X0 Stainless Steel Plate (50 mm) is an upper plate compatible with lower Standard and Advanced Peltier plates. It features Smart Swap 2 geometry for accurate placement and consistent measurement on HR X0 instruments.
Catalog No. 50-005-80782 Supplier TA Instruments Supplier No. 511500.945
HR X0 Stainless Steel Plate (50 mm) is an upper plate compatible with lower Standard and Advanced Peltier plates. It features Smart Swap 2 geometry for accurate placement and consistent measurement on HR X0 instruments.