Learn More
Alkali Scientific Stainless Steel 8 High Frame for C 50 Canisters
SDP
Catalog No. 5000662021
Encompass_Preferred
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 8 High Frame for C 50 Holds 8 C 50 Canisters Blood Bag Number: BAXTER/FENWAL 4R9951, OrigenCS50, CyroMACS 200 074 400, CharterMed CF 50, FP 50mL and FP FLEX50 Dimensions (D x H x L): 5 11/16 x 29 3/4 x 9/16″ 145 x 755 x 15mm Canister: C 50
Catalog No. 50-006-62021 Supplier Alkali Scientific Supplier No. FC508
Universal Canister Frames are constructed of corrosion resistant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. 8 High Frame for C 50 Holds 8 C 50 Canisters Blood Bag Number: BAXTER/FENWAL 4R9951, OrigenCS50, CyroMACS 200 074 400, CharterMed CF 50, FP 50mL and FP FLEX50 Dimensions (D x H x L): 5 11/16 x 29 3/4 x 9/16″ 145 x 755 x 15mm Canister: C 50