Learn More
Alkali Scientific Alkali Scientific™ Stainless Steel (Acid-Washed) beads, 2.8mm, Bulk 1000/Pk
SDP
Catalog No. 503027505
Encompass_Preferred
Stainless Steel (Acid-Washed) beads, 2.8mm, Bulk 1000/Pk   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.    
Catalog No. 50-302-7505 Supplier Alkali Scientific Supplier No. RS70133
Stainless Steel (Acid-Washed) beads, 2.8mm, Bulk 1000/Pk   Compatible with BeadBug™ Microtube homogenizer RS7020, BeadBlaster™ Microtube homogenizer D8400.    