Learn More
Analytical Sales & Services Stainless Steel Adjustable Flow Splitter with -03 resistor, Post Column, Ratio 15:1 to 250:1
Catalog No. 5000806515
Encompass
Advantage Model 62 Stainless Steel Adjustable Flow Splitter with -03 resistor 15:1 to 250:1 Post Column Analytical Range\n\nQty/Pkg: EACH
Catalog No. 50-008-06515 Supplier Analytical Sales & Services Supplier No. 62412
Advantage Model 62 Stainless Steel Adjustable Flow Splitter with -03 resistor 15:1 to 250:1 Post Column Analytical Range\n\nQty/Pkg: EACH