Learn More
Analytical Sales & Services Stainless Steel Adjustable Flow Splitter with -04 resistor, Post Column, Ratio 5:1 to 100:1
Catalog No. 5000806345
Encompass
Advantage Model 62 Stainless Steel Adjustable Flow Splitter with -04 resistor 5:1 to 100:1 Post Column Splitter Analytical Range\n\nQty/Pkg: EACH
Catalog No. 50-008-06345 Supplier Analytical Sales & Services Supplier No. 62413
Advantage Model 62 Stainless Steel Adjustable Flow Splitter with -04 resistor 5:1 to 100:1 Post Column Splitter Analytical Range\n\nQty/Pkg: EACH