Learn More
TA Instruments Stainless Steel Bail Pkg of 15, for Use with Aluminum Sample Pans P/N 957363.901, Aluminum Sample Lids P/N 957362. 901
Catalog No. 5000580890 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Bail Pkg Of 15 is used with the aluminum sample pans P/N 957363.901 and the aluminum sample lids P/N 957362.901.
Catalog No. 50-005-80890 Supplier TA Instruments Supplier No. 957364.901
Waters TA Instruments Stainless Steel Bail Pkg Of 15 is used with the aluminum sample pans P/N 957363.901 and the aluminum sample lids P/N 957362.901.