Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DR Instruments Stainless Steel Bandage Scissors, 5.5 in.
SDPSCI_ED

Catalog No. S20674
Change view
Click to view available options
Length (English):
5.5 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S20674 5.5 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S20674 Supplier DR Instruments Supplier No. 5BND
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stainless Steel Bandage Scissors to size bandages and dressings and cut through medical gauze.

  • One blunt tip and one spatula tip for easier cutting
  • Precision ground blades for cleaner and easier cutting
  • Silver polished finish
  • 5.5 in. Length

Specifications

Product Type Bandage Scissors
Length (English) 5.5 in.
Length (Metric) 14 cm
Material Stainless Steel
Color Silver Polished Finish
Description One blunt tip/one spatula tip for easy cutting
Finish Silver Polished
For Use With (Application) To size bandages, dressings and cut through medical gauze
Tip Style Blunt Point, Spatula Point
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.