Learn More
Brady™ Banding Heads
Catalog No. 19111925 Shop All Brady Corporation Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-111-925 1 Ea.
1 options
Catalog No. 19-111-925 Supplier Brady™ Supplier No. 90910
  • $110.00
    $86.65 / Pack of 100
    Save $23.35
    21% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Used for creating secure closure. Brady™ Banding Heads fits 1/4 in. strapping and is made up of Stainless Steel material.
  • Heads are made of Stainless Steel material
  • Use with Continuous Strapping to create secure closures
  • Fits 1/4 in. strapping
Specifications
Material Stainless Steel (316)
Quantity 1 Ea.
WARNING: Cancer - www.P65Warnings.ca.gov