Learn More
Elma™ Stainless Steel Basket for Elmasonic S50R Ultrasonic Sieve Cleaner
SDP
Catalog No. 08000138
Change view
Click to view available options
Diameter (Metric) Inner:
220 mm
Catalog No. Diameter (Metric) Inner
08-000-138 220 mm
1 options
Catalog No. 08-000-138 Supplier Elma™ Supplier No. 104 6006
Add to Cart
Add to Cart
For use with Elmasonic S50R Ultrasonic Sieve Cleaner.
  • For laboratory vessels and other items
  • Open weave design
Specifications
Product Type Basket
Diameter (Metric) Inner 220 mm
For Use With (Equipment) Ultrasonic Cleaner, Elmasonic S50R
For Use With (Application) Hold Parts When Immersing and Removing Them From Solution in Ultrasonic Cleaners
Material Stainless Steel