Learn More
Elma™ Stainless Steel Baskets for Elmasonic Xtra ST Ultrasonic Cleaner
Description
- Holds parts when immersing and removing from solution
- Open weave in sides and bottoms
- Protect cleaner and parts from damage caused by direct contact
Specifications
Specifications
| Product Type | Basket |
| For Use With (Equipment) | Ultrasonic Cleaner, Elmasonic Xtra ST 300 H |
| For Use With (Application) | Hold Parts When Immersing and Removing Them From Solution in Ultrasonic Cleaners |
| Material | Stainless Steel |
| Dimensions (L x W x H) | 24.5 x 22.8 x 17 cm |