Learn More
Cole-Parmer Cole-Parmer Stainless steel beaker, 1.2 L
Supplier:  Cole-Parmer 0720610
Encompass_Preferred
These Cole-Parmer beakers are constructed of durable 304 medium gauge stainless steel. Recessed covers are ideal for maximizing storage space. Cover must be purchased separately.
Catalog No. NC0364534