Learn More
BURKLE INC Stainless steel beaker, extended, 33.81 oz (1000 ml), TeleScoop
Catalog No. 5000025575
Encompass_Preferred
Sampling dipper made of stainless steel AISI 304 (1.4301) with extended, completely closed handle.
The sample beaker has no openings or indentations to an immersion depth of approx. 500 mm (19.69 in.). The closed design of the rod makes the scoop ideal for sampling water from wells, sludge, wastewater or liquid manure.
In combination with the telescopic rod, a range of up to 6 metres (19.69 ft.) is possible.
Thanks to the scraper edge on the rim of the beaker, samples can also be taken of viscous media, powders and granulates.
The beaker is steam-sterilizable and completely flame-resistant, which makes it easy to sterilize at the point of use.
Catalog No. 50-000-25575 Supplier BURKLE INC Supplier No. 53542010
Add to Cart
Add to Cart
Sampling dipper made of stainless steel AISI 304 (1.4301) with extended, completely closed handle.
The sample beaker has no openings or indentations to an immersion depth of approx. 500 mm (19.69 in.). The closed design of the rod makes the scoop ideal for sampling water from wells, sludge, wastewater or liquid manure.
In combination with the telescopic rod, a range of up to 6 metres (19.69 ft.) is possible.
Thanks to the scraper edge on the rim of the beaker, samples can also be taken of viscous media, powders and granulates.
The beaker is steam-sterilizable and completely flame-resistant, which makes it easy to sterilize at the point of use.