Learn More
Electron Microscopy Sciences Stainless Steel. Biopsy Insert
SDP
Catalog No. 5029857
Encompass_Preferred
A two-part loose fitting stainless steel capsule with welded mesh that fits inside the processing cassettes preventing the loss of small or friable material while allowing interchange of fluids. They fit all process/embedding cassettes such as 70025 and 72025 as well as Uni-Cassettes (62500).
Catalog No. 50-298-57 Supplier Electron Microscopy Sciences Supplier No. 6232801
A two-part loose fitting stainless steel capsule with welded mesh that fits inside the processing cassettes preventing the loss of small or friable material while allowing interchange of fluids. They fit all process/embedding cassettes such as 70025 and 72025 as well as Uni-Cassettes (62500).