Learn More
TA Instruments Stainless Steel Blank Plug for Peltier Double Gap Cup, Converts Double Gap Cup to Large Diameter Cup 44 mm, Compatible with All Upper Rotors, for HR X0, DHR and AR-Series Ver 1 Cups
Catalog No. 5000580808 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Blank Plug For Peltier Double Gap Cup is for use with HR X0, DHR, and AR-Series Peltier Concentric Cylinder Double Gap Cups (Ver 1). This plug converts a double gap cup into a large diameter cup with a diameter of 44 mm. The cup is compatible with all upper rotors, and sample volume depends on rotor selection.
Catalog No. 50-005-80808 Supplier TA Instruments Supplier No. 545642.001
Waters TA Instruments Stainless Steel Blank Plug For Peltier Double Gap Cup is for use with HR X0, DHR, and AR-Series Peltier Concentric Cylinder Double Gap Cups (Ver 1). This plug converts a double gap cup into a large diameter cup with a diameter of 44 mm. The cup is compatible with all upper rotors, and sample volume depends on rotor selection.