Learn More
Braintree Scientific Stainless Steel Brain Matrice,Mouse 40-75gm, 0.5 mm Coronal
SDP
Catalog No. 501954417
Encompass
Stainless Steel Brain Matrices These highly polished high quality stainless steel matrices are precisely machined to ensure reproducible sections. The brain matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1mm or 0.5mm intervals. **Sold for research purposes only and not for human use**
Catalog No. 50-195-4417 Supplier Braintree Scientific Supplier No. BSSS505C
 Restricted Item
Add to Cart
Add to Cart
Stainless Steel Brain Matrices These highly polished high quality stainless steel matrices are precisely machined to ensure reproducible sections. The brain matrices allow the investigator to slice either coronal (perpendicular to center line) or sagittal (parallel to center line) sections through the brain at 1mm or 0.5mm intervals. **Sold for research purposes only and not for human use**