Learn More
World Precision Instrument Stainless Steel Brain Matrix, Adult mouse, Cor. Slices, 40-75g
SDP
Catalog No. 502538600
Encompass
Using a WPI brain matrix, you can get thin and uniform brain slices, which is crucial for histological analysis, immunohistochemistry, molecular biology, and more. This consistency is essential for accurate comparisons between different brains and experimental conditions. These WPI rodent brain matrices are made of stainless steel, a durable material. They have precision-cut channels or slots that hold the brain in place during slicing. Choose your brain matrix based on your species (mouse or rat) and the size of the animal. The WPI brain matrices are designed for sagittal or coronal slices. These brain matrices give you consistent slices that are 1 mm thick, and the olfactory spinal notch is cut into every brain matrix. Matrices may be chilled and sterilized. These are useful when heat transfer or maintaining a specific temperature range is required. Get standardized and precise brain sectioning.
Catalog No. 50-253-8600 Supplier World Precision Instrument Supplier No. RBMS200C
Using a WPI brain matrix, you can get thin and uniform brain slices, which is crucial for histological analysis, immunohistochemistry, molecular biology, and more. This consistency is essential for accurate comparisons between different brains and experimental conditions. These WPI rodent brain matrices are made of stainless steel, a durable material. They have precision-cut channels or slots that hold the brain in place during slicing. Choose your brain matrix based on your species (mouse or rat) and the size of the animal. The WPI brain matrices are designed for sagittal or coronal slices. These brain matrices give you consistent slices that are 1 mm thick, and the olfactory spinal notch is cut into every brain matrix. Matrices may be chilled and sterilized. These are useful when heat transfer or maintaining a specific temperature range is required. Get standardized and precise brain sectioning.