Learn More
Strem, An Ascensus Company 1cyl. Stainless steel bubbler, 150ml, vertical, electropolished with fill-port, PCTFE valve stem tip, DOT 4B, UN stamped.
SDP
Catalog No. 502045438
Encompass
1cyl. Stainless steel bubbler, 150ml, vertical, electropolished with fill-port, PCTFE valve stem tip, DOT 4B, UN stamped. . Molecular Weight: . Molecular Formula: . Color/form: Strem# 95-4151. http://www.strem.com/catalog/v/95-4151/
Catalog No. 50-204-5438 Supplier Strem, An Ascensus Company Supplier No. 9541511CYL
1cyl. Stainless steel bubbler, 150ml, vertical, electropolished with fill-port, PCTFE valve stem tip, DOT 4B, UN stamped. . Molecular Weight: . Molecular Formula: . Color/form: Strem# 95-4151. http://www.strem.com/catalog/v/95-4151/