Learn More
Strem, An Ascensus Company 1cyl. Stainless steel bubbler, 600ml, vertical, electropolished with fill-port, pneumatic actuator valves, DOT 4B.
SDP
Catalog No. 502065541
Encompass
1cyl. Stainless steel bubbler, 600ml, vertical, electropolished with fill-port, pneumatic actuator valves, DOT 4B. . Molecular Weight: . Molecular Formula: . Color/form: Strem# 95-1050. http://www.strem.com/catalog/v/95-1050/
Catalog No. 50-206-5541 Supplier Strem, An Ascensus Company Supplier No. 9510501CYL
Add to Cart
Add to Cart
Request a Quote
1cyl. Stainless steel bubbler, 600ml, vertical, electropolished with fill-port, pneumatic actuator valves, DOT 4B. . Molecular Weight: . Molecular Formula: . Color/form: Strem# 95-1050. http://www.strem.com/catalog/v/95-1050/