Learn More
Sigma Aldrich Fine Chemicals Biosciences Stainless steel bubbler ca1EA
Catalog No. 502908780
Encompass
Valve ParametersFlow coefficient (Cv) 0.29 XT 0.6Operating temperature -40 to 160F (-40 to 71C)Surface finish 15 uin. Ra maxInboard leakage 2 x 10-10 SCCSOutboard leakage 2 x 10-9 SCCS He at 250 psig inlet pressureLeakage across seat 4 x 10-8 SCCS He at 250 psig inlet pressure
Catalog No. 50-290-8780 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z5533601EA
Valve ParametersFlow coefficient (Cv) 0.29 XT 0.6Operating temperature -40 to 160F (-40 to 71C)Surface finish 15 uin. Ra maxInboard leakage 2 x 10-10 SCCSOutboard leakage 2 x 10-9 SCCS He at 250 psig inlet pressureLeakage across seat 4 x 10-8 SCCS He at 250 psig inlet pressure