Learn More
TA Instruments Stainless Steel Building Materials Cell Cup with Slotted Cage, Diameter 57 mm, Height 75 mm, 26 Slots, Width 5 mm, Length 67 mm, Thickness 1.5 mm, for Building Materials Paddle P/N 533247.901
Catalog No. 5000580836 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Stainless Steel Building Materials Cell Cup With Slotted Cage is for use with the Building Materials Paddle (P/N 533247.901) for characterization of cements and mortars. The cup includes a removable slotted cage to minimize sample slippage. The cage has 26 equally spaced slots with width 5 mm, length 67 mm, and thickness 1.5 mm. Cup dimensions: diameter 57 mm, height 75 mm.
Catalog No. 50-005-80836 Supplier TA Instruments Supplier No. 533246.901
Waters TA Instruments Stainless Steel Building Materials Cell Cup With Slotted Cage is for use with the Building Materials Paddle (P/N 533247.901) for characterization of cements and mortars. The cup includes a removable slotted cage to minimize sample slippage. The cage has 26 equally spaced slots with width 5 mm, length 67 mm, and thickness 1.5 mm. Cup dimensions: diameter 57 mm, height 75 mm.