Learn More
TA Instruments Stainless Steel Building Materials Cell Paddle, Width 44 mm, Height 30 mm, Two Slots Width 15 mm, Height 26 mm, for Cell Cup P/N 533246.901
Supplier:  TA Instruments 533247.901
Encompass_Preferred
Waters TA Instruments Stainless Steel Building Materials Cell Paddle is for use with the Building Materials Cell Cup (P/N 533246.901) for characterization of cements and mortars. The paddle is 44 mm wide and 30 mm high. It has two slots to promote sample mixing, each with width 15 mm and height 26 mm.
Catalog No. 50-005-80758