Learn More
Fine Science Tools Burrs for Micro Drill 0.9mm
SDP
Catalog No. NC9523897
Encompass
Tip Diameter: 0.9mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Stainless Steel/ Length: 44mm/ Autoclave Safe: Yes
Catalog No. NC9523897 Supplier Fine Science Tools Supplier No. 1900809
Tip Diameter: 0.9mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Stainless Steel/ Length: 44mm/ Autoclave Safe: Yes