Learn More
Alkali Scientific Stainless Steel Canister Frames for BAXTER/FENWAL Blood Bag # 4R 2110 (4 Canisters)
SDP
Catalog No. 5000662042
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 4 BAXTER/FENWAL C 4R2110 Aluminum Canisters Blood Bag Number: 4R 2110 Dimensions (D x H x L): 9 3/16 x 33 1/2 x 15/16″ 234 x 851 x 24mm Canister: C 4R2110 Platform Dividers P20 4R2110 P24 4R2110 P31 4R2110 P39 4R2110
Catalog No. 50-006-62042 Supplier Alkali Scientific Supplier No. FC4R21104
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 4 BAXTER/FENWAL C 4R2110 Aluminum Canisters Blood Bag Number: 4R 2110 Dimensions (D x H x L): 9 3/16 x 33 1/2 x 15/16″ 234 x 851 x 24mm Canister: C 4R2110 Platform Dividers P20 4R2110 P24 4R2110 P31 4R2110 P39 4R2110