Learn More
Alkali Scientific Stainless Steel Canister Frames for BAXTER/FENWAL Blood Bag # 4R 2111 (4 Canisters)
SDP
Catalog No. 5000662044
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 4 BAXTER/FENWAL C 4R2111 Aluminum Canisters Blood Bag Number: 4R 2111 Dimensions (D x H x L): 7 11/16 x 26 9/16 x 15/16″ 196 x 674 x 24mm Canister: C 4R2111 Platform Dividers P20 4R2111 P24 4R2111 P31 4R2111 P39 4R2111
Catalog No. 50-006-62044 Supplier Alkali Scientific Supplier No. FC4R21114
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 4 BAXTER/FENWAL C 4R2111 Aluminum Canisters Blood Bag Number: 4R 2111 Dimensions (D x H x L): 7 11/16 x 26 9/16 x 15/16″ 196 x 674 x 24mm Canister: C 4R2111 Platform Dividers P20 4R2111 P24 4R2111 P31 4R2111 P39 4R2111