Learn More
Alkali Scientific Stainless Steel Canister Frames for BAXTER/FENWAL Blood Bag # 4R 9953 & 4R 5461 (3 Canisters)
SDP
Catalog No. 5000662314
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 3 BAXTER/FENWAL C 4R9953 Aluminum Canisters Blood Bag Number: 4R 9953 4R 5461 Dimensions (D x H x L): 7 5/16 x 17 5/16 x 9/16″ 186 x 440 x 15mm Canister: C 4R9953 Platform Dividers P20 4R9953 P24 4R9953 P31 4R9953 P39 4R9953
Catalog No. 50-006-62314 Supplier Alkali Scientific Supplier No. FC4R99533
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 3 BAXTER/FENWAL C 4R9953 Aluminum Canisters Blood Bag Number: 4R 9953 4R 5461 Dimensions (D x H x L): 7 5/16 x 17 5/16 x 9/16″ 186 x 440 x 15mm Canister: C 4R9953 Platform Dividers P20 4R9953 P24 4R9953 P31 4R9953 P39 4R9953