Learn More
Alkali Scientific Stainless Steel Canister Frames for BAXTER/FENWAL Blood Bag # 4R 9957 & 4R 5463 (3 Canisters)
Supplier: Alkali Scientific FC4R99573
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frame for 3 BAXTER/FENWAL C 4R9957 Aluminum Canisters Blood Bag Number: 4R 9957 4R 5463 Dimensions (D x H x L): 10 5/16 x 17 5/16 x 9/16″ 262 x 440 x 15mm Canister: C 4R9957 Platform Dividers P20 4R9957 P24 4R9957 P31 4R9957 P39 4R9957