Learn More
Alkali Scientific Stainless Steel Canister Frames for BAXTER/FENWAL Blood Bag # 4R 9957 & 4R 5463 (5 Canisters)
SDP
Catalog No. 5000662303
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 5 BAXTER/FENWAL C 4R9957 Aluminum Canisters Blood Bag Number: 4R 9957 4R 5463 Dimensions (D x H x L): 10 5/16 x 28 9/16 x 9/16″ 262 x 726 x 15mm Canister: C 4R9957 Platform Dividers P20 4R9957 P24 4R9957 P31 4R9957 P39 4R9957
Catalog No. 50-006-62303 Supplier Alkali Scientific Supplier No. FC4R99575
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 5 BAXTER/FENWAL C 4R9957 Aluminum Canisters Blood Bag Number: 4R 9957 4R 5463 Dimensions (D x H x L): 10 5/16 x 28 9/16 x 9/16″ 262 x 726 x 15mm Canister: C 4R9957 Platform Dividers P20 4R9957 P24 4R9957 P31 4R9957 P39 4R9957