Learn More
Alkali Scientific Stainless Steel Canister Frames for BAXTER/FENWAL Blood Bag # 4R 9959 & 4R 5464 (5 Canisters)
SDP
Catalog No. 5000662981
Encompass_Preferred
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 5 BAXTER/FENWAL C 4R9959 Aluminum Canisters Blood Bag Number: 4R 9959 4R 5464 Dimensions (D x H x L): 12 11/16 x 28 9/16 x 9/16″ 322 x 726 x15mm Canister: C 4R9959 Platform Dividers P20 4R9959 P24 4R9959 P31 4R9959 P39 4R9959
Catalog No. 50-006-62981 Supplier Alkali Scientific Supplier No. FC4R99595
Canister Frames are constructed of corrosion restant stainless steel. Each Frame is designed to hold a specific Canister and includes a handle. Stainless Steel Frames for 5 BAXTER/FENWAL C 4R9959 Aluminum Canisters Blood Bag Number: 4R 9959 4R 5464 Dimensions (D x H x L): 12 11/16 x 28 9/16 x 9/16″ 322 x 726 x15mm Canister: C 4R9959 Platform Dividers P20 4R9959 P24 4R9959 P31 4R9959 P39 4R9959